Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci VTI1a Peptide
SDP

Catalog No. 16595824
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-824 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-824 Supplier ProSci Supplier No. 8287P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

VTI1a peptide is used for blocking the activity of VTI1a antibody.

Specifications

Gene ID (Entrez) 143187
Species Human
Accession Number NP_660207
Common Name vesicle transport through interaction with t-SNAREs 1 A
Concentration 200 μg/mL
Content And Storage Store VTI1a peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol VTI1a
Quantity 0.05 mg
Sequence 19 amino acid peptide near the center of human VTI1a.
Synonym Vesicle transport through interaction with t-SNAREs 1 A, MMDS3, MVti1, VTI1RP2, Vti1-rp2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.