Learn More
Aviva Systems Biology Corp VTI1B Antibody - C-terminal region (ARP61763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VTI1B. It was validated on Western Blot. . Alternate names: VTI1, VTI1-LIKE, VTI1L, VTI2, v-SNARE, vti1-rp1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-461