Learn More
Alpco Ltd Vitamin B6 HPLC
SDP
Supplier:  Alpco Ltd 302100
Encompass
Vitamin B6 is the term for pyridoxin, pyridoxal and pyridoxamin and the appropriate phosphate products. All forms can be transformed into the active form pyridoxal-5-phosphate. Vitamin B6 functions as a coenzyme and is essential for more than 50 reactions in the protein metabolism thereby synthesizing, transforming or degrading amino acids. Vitamin B6 supports the resorption of amino acids and their transport into the cells. Furthermore, vitamin B6 contributes to the synthesis of neurotransmitters and amine products (histamine).
Catalog No. NC1678919