Learn More
Aviva Systems Biology Corp VTN Antibody - N-terminal region (ARP58095_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58095_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VTN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VTN. Alternate names: V75, VN, VNTApplicationWB. Species reactivity: Human
Catalog No. 50-014-62236