Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VTP-27999 Hydrochloride | 1264191-73-2 | 98.2% | C26H42Cl2N4O5 | 10MM/1ML
SDP

Catalog No. 5000434477
Encompass_Preferred

VTP-27999 Hcl is an alkyl amine Renin inhibitor that is useful for Hypertension and End-Organ Diseases. Its IC50 value targets Renin.

  • Molecular weight: 561.54
  • Appearance: Solid
  • Color: White to yellow
  • Solubility in water: ≥ 100 mg/mL (178.08 mM)
  • Storage as solid: 4°C, sealed, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-34477 Supplier Medchemexpress LLC Supplier No. HY7665210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VTP-27999 Hcl is an alkyl amine Renin inhibitor that is useful for Hypertension and End-Organ Diseases. Its IC50 value targets Renin.

  • Molecular weight: 561.54
  • Appearance: Solid
  • Color: White to yellow
  • Solubility in water: ≥ 100 mg/mL (178.08 mM)
  • Storage as solid: 4°C, sealed, away from moisture
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.