Learn More
Medchemexpress LLC VTP-27999 (TFA) | 1013937-63-7 | 97.0% | C28H42ClF3N4O7 | 5 MG
SDP
Catalog No. 5000358084
Encompass_Preferred
VTP-27999 (TFA) is an alkyl amine Renin inhibitor useful for hypertension and end-organ diseases. This product is for research use only.
  • Alkyl amine Renin inhibitor
  • Useful for hypertension research
  • Useful for end-organ diseases research
  • For research use only
Catalog No. 50-003-58084 Supplier Medchemexpress LLC Supplier No. HY507695MG
VTP-27999 (TFA) is an alkyl amine Renin inhibitor useful for hypertension and end-organ diseases. This product is for research use only.
  • Alkyl amine Renin inhibitor
  • Useful for hypertension research
  • Useful for end-organ diseases research
  • For research use only