Learn More
Medchemexpress LLC VTP-27999 TFA | 1013937-63-7 | 97.00% | C28H42ClF3N4O7 | 25 MG
SDP
Catalog No. 5000359119
Encompass_Preferred
VTP-27999 TFA is a potent alkyl amine Renin inhibitor, recognized for its role in addressing hypertension and related end-organ diseases. This compound is primarily utilized in research settings to explore the mechanisms of renin-angiotensin-aldosterone system (RAAS) regulation. Its specific inhibitory action offers a valuable tool for studying cardiovascular conditions and developing novel therapeutic strategies.
  • Potent alkyl amine renin inhibitor
  • Useful for hypertension research
  • Applicable in studies of end-organ diseases
  • Supports research on RAAS regulation
  • Aids in the development of cardiovascular therapies
Catalog No. 50-003-59119 Supplier Medchemexpress LLC Supplier No. 25MGHY50769
VTP-27999 TFA is a potent alkyl amine Renin inhibitor, recognized for its role in addressing hypertension and related end-organ diseases. This compound is primarily utilized in research settings to explore the mechanisms of renin-angiotensin-aldosterone system (RAAS) regulation. Its specific inhibitory action offers a valuable tool for studying cardiovascular conditions and developing novel therapeutic strategies.
  • Potent alkyl amine renin inhibitor
  • Useful for hypertension research
  • Applicable in studies of end-organ diseases
  • Supports research on RAAS regulation
  • Aids in the development of cardiovascular therapies