Learn More
Medchemexpress LLC VTP50469
SDP
Catalog No. NC3398158
Encompass_Preferred
Solid + Solvent. Ready for reconstitution. VTP50469 is a potent, highly selective, and orally active Menin-MLL interaction inhibitor with a Ki of 104 pM. VTP50469 has potently anti-leukemia activity.
For research use only. We do not sell to patients.
Catalog No. NC3398158 Supplier Medchemexpress LLC Supplier No. HY114162
Solid + Solvent. Ready for reconstitution. VTP50469 is a potent, highly selective, and orally active Menin-MLL interaction inhibitor with a Ki of 104 pM. VTP50469 has potently anti-leukemia activity.
For research use only. We do not sell to patients.