Learn More
Selleck Chemical LLC VTP50469 5mg
SDP
Supplier:  Selleck Chemical LLC S89345MG
Encompass_Preferred
VTP50469 is a potent, highly selective and orally active inhibitor of Menin-MLL interaction with Ki of 104 pM. VTP50469 exhibits anti-leukemia activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3444