Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VTP50469 fumarate | 2169919-29-1 | 99.79% | 804.93 | 10 MG
SDP

Catalog No. 5000437856
Encompass_Preferred

VTP50469 fumarate | 2169919-29-1 | 99.79% | 804.93 | 10 MG

Catalog No. 50-004-37856 Supplier Medchemexpress LLC Supplier No. HY114162A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VTP50469 fumarate | 2169919-29-1 | 99.79% | 804.93 | 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.