Learn More
Medchemexpress LLC VTP50469 fumarate | 2169919-29-1 | 99.8% | 804.93 | 100 MG
SDP
Catalog No. 5000435814
Encompass_Preferred
VTP50469 fumarate is a potent, highly selective, and orally active Menin-MLL interaction inhibitor. It demonstrates potent anti-leukemia activity with a Ki of 104 pM.
  • Inhibits cell proliferation in MLL-r and ALL cell lines
  • Induces apoptosis in MLL-r B cell ALL cell lines
  • Causes dose-dependent differentiation in MLL-r AML cell lines
  • Displaces Menin from protein complexes and inhibits chromatin occupancy of MLL
  • Leads to changes in gene expression, differentiation, and apoptosis
  • Shows significant survival advantage in in vivo studies in NSG mice
Catalog No. 50-004-35814 Supplier Medchemexpress LLC Supplier No. HY114162A100MG
VTP50469 fumarate is a potent, highly selective, and orally active Menin-MLL interaction inhibitor. It demonstrates potent anti-leukemia activity with a Ki of 104 pM.
  • Inhibits cell proliferation in MLL-r and ALL cell lines
  • Induces apoptosis in MLL-r B cell ALL cell lines
  • Causes dose-dependent differentiation in MLL-r AML cell lines
  • Displaces Menin from protein complexes and inhibits chromatin occupancy of MLL
  • Leads to changes in gene expression, differentiation, and apoptosis
  • Shows significant survival advantage in in vivo studies in NSG mice