Learn More
Medchemexpress LLC VTP50469 fumarate | 2169919-29-1 | 99.8% | 804.93 g/mol | C32H47FN6O4S · 3/2 C4H4O4 | 5 MG
SDP
Catalog No. 5000262063
Encompass_Preferred
VTP50469 fumarate is the fumarate salt of VTP50469, a potent, highly selective, orally active inhibitor of the Menin-MLL (MLL1) protein-protein interaction. It has a reported Ki of 104 pM and demonstrates potent anti-leukemia activity in preclinical studies. The compound is supplied for research use with supporting datasheet, certificate of analysis, and safety data.
  • Highly potent inhibitor with a reported Ki of 104 pM.
  • Orally active compound effective in preclinical leukemia models.
  • Selective mode of action targeting the Menin-MLL interaction.
  • High purity suitable for biochemical and cellular studies.
  • Supplied with datasheet, COA, and SDS for lab use.
  • Available in small research quantities for preclinical experiments.
Catalog No. 50-002-62063 Supplier Medchemexpress LLC Supplier No. HY114162A5MG
VTP50469 fumarate is the fumarate salt of VTP50469, a potent, highly selective, orally active inhibitor of the Menin-MLL (MLL1) protein-protein interaction. It has a reported Ki of 104 pM and demonstrates potent anti-leukemia activity in preclinical studies. The compound is supplied for research use with supporting datasheet, certificate of analysis, and safety data.
  • Highly potent inhibitor with a reported Ki of 104 pM.
  • Orally active compound effective in preclinical leukemia models.
  • Selective mode of action targeting the Menin-MLL interaction.
  • High purity suitable for biochemical and cellular studies.
  • Supplied with datasheet, COA, and SDS for lab use.
  • Available in small research quantities for preclinical experiments.