Learn More
Medchemexpress LLC HY-112782 2mg , VTX-27 CAS:1321924-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127822MG
Encompass_Preferred
HY-112782 2mg VTX-27 CAS: 1321924-70-2 VTX-27 is a selective protein kinase C θ (PKC θ) inhibitor, with Kis of 0.08 nM and 16 nM for PKC θ and PKC δ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9949