Learn More
Medchemexpress LLC Vu 0238429 | 1160247-92-6 | 99.9% | 100 MG
SDP
Catalog No. 5000232423
Encompass_Preferred
VU 0238429 is a selective positive allosteric modulator of the muscarinic acetylcholine receptor subtype 5 (mAChR5, M5) used as a research tool to probe M5 receptor pharmacology.
  • Positive allosteric modulator of mAChR5 with EC50 1.16 μM.
  • High purity (≈99.9%), suitable for research applications.
  • Solid, light yellow to orange appearance.
  • Stable under recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Relevant to GPCR/G protein and neuronal signaling studies.
Catalog No. 50-002-32423 Supplier Medchemexpress LLC Supplier No. HY12157100MG
VU 0238429 is a selective positive allosteric modulator of the muscarinic acetylcholine receptor subtype 5 (mAChR5, M5) used as a research tool to probe M5 receptor pharmacology.
  • Positive allosteric modulator of mAChR5 with EC50 1.16 μM.
  • High purity (≈99.9%), suitable for research applications.
  • Solid, light yellow to orange appearance.
  • Stable under recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Relevant to GPCR/G protein and neuronal signaling studies.