Learn More
TARGETMOL CHEMICALS INC VU 0238429 25MG
SDP
Catalog No. 502879738
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. VU 0238429 (SCR-1481B1) is positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5 or M5)( EC50 1.16 uM). Purity 100%
Catalog No. 50-287-9738 Supplier TARGETMOL CHEMICALS INC Supplier No. T800725MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. VU 0238429 (SCR-1481B1) is positive allosteric modulator of muscarinic acetylcholine receptor subtype 5 (mAChR5 or M5)( EC50 1.16 uM). Purity 100%