Learn More
Medchemexpress LLC VU 0240551 | 893990-34-6 | 99.7% | 342.44 | 100 MG
SDP
Catalog No. 5000392430
Encompass_Preferred
VU 0240551 is a potent neuronal K-Cl cotransporter KCC2 inhibitor (IC50=560 nM) that is selective versus NKCC1. It also inhibits hERG and L-type Ca2+ channels, making it a valuable tool in neuroscience research. This compound attenuates GABA-induced hyperpolarization of P cells, produces a positive shift in the P cell GABA reversal potential, and enhances P cell synaptic transmission.
  • Potent neuronal K-Cl cotransporter KCC2 inhibitor
  • Selective inhibition over NKCC1
  • Inhibits hERG and L-type Ca2+ channels
  • Attenuates GABA-induced hyperpolarization of P cells
  • Enhances P cell synaptic transmission
Catalog No. 50-003-92430 Supplier Medchemexpress LLC Supplier No. HY16689100MG
VU 0240551 is a potent neuronal K-Cl cotransporter KCC2 inhibitor (IC50=560 nM) that is selective versus NKCC1. It also inhibits hERG and L-type Ca2+ channels, making it a valuable tool in neuroscience research. This compound attenuates GABA-induced hyperpolarization of P cells, produces a positive shift in the P cell GABA reversal potential, and enhances P cell synaptic transmission.
  • Potent neuronal K-Cl cotransporter KCC2 inhibitor
  • Selective inhibition over NKCC1
  • Inhibits hERG and L-type Ca2+ channels
  • Attenuates GABA-induced hyperpolarization of P cells
  • Enhances P cell synaptic transmission