Learn More
Medchemexpress LLC Vu 0255035 | 1135243-19-4 | 99.8% | 432.52 | C18H20N6O3S2 | 5 MG
SDP
Catalog No. 5000242499
Encompass_Preferred
VU 0255035 is a selective M1 muscarinic receptor antagonist used as a research tool to probe M1-mediated signaling and synaptic responses in neuronal preparations. It is supplied as a solid with high purity and is characterized by its molecular formula and weight; handle under recommended storage conditions for best stability.
  • Selective M1 muscarinic receptor antagonist with demonstrated neuronal activity.
  • High purity: 99.8%.
  • Molecular weight 432.52; formula C18H20N6O3S2.
  • Available in small research quantities, such as 5 mg.
  • Supplied as a solid; store at 4°C and protect from light (in solvent: -80°C for long-term storage).
  • Suitable as a pharmacological probe for electrophysiology and signaling assays.
Catalog No. 50-002-42499 Supplier Medchemexpress LLC Supplier No. HY1082345MG
VU 0255035 is a selective M1 muscarinic receptor antagonist used as a research tool to probe M1-mediated signaling and synaptic responses in neuronal preparations. It is supplied as a solid with high purity and is characterized by its molecular formula and weight; handle under recommended storage conditions for best stability.
  • Selective M1 muscarinic receptor antagonist with demonstrated neuronal activity.
  • High purity: 99.8%.
  • Molecular weight 432.52; formula C18H20N6O3S2.
  • Available in small research quantities, such as 5 mg.
  • Supplied as a solid; store at 4°C and protect from light (in solvent: -80°C for long-term storage).
  • Suitable as a pharmacological probe for electrophysiology and signaling assays.