Learn More
Medchemexpress LLC Vu 0255035 | 1135243-19-4 | 99.8% | 432.52 g·mol⁻¹ | C18H20N6O3S2 | 1 ML
SDP
Catalog No. 5000243393
Encompass_Preferred
VU 0255035 is a selective, competitive antagonist of the muscarinic M1 receptor supplied for research use. It is available as a high-purity solid and as a ready-to-use 10 mM solution in DMSO, supporting receptor pharmacology experiments, selectivity profiling, and dose-response studies.
Catalog No. 50-002-43393 Supplier Medchemexpress LLC Supplier No. HY10823410MM 1ML
VU 0255035 is a selective, competitive antagonist of the muscarinic M1 receptor supplied for research use. It is available as a high-purity solid and as a ready-to-use 10 mM solution in DMSO, supporting receptor pharmacology experiments, selectivity profiling, and dose-response studies.