Learn More
TARGETMOL CHEMICALS INC VU 0285683 5MG
SDP
Catalog No. 502580865
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU 0285683 is a specific negative allosteric modulator of mGluR5 with a high affinity for the MPEP binding site. VU 0285683 shows anxiolytic-like activity. purity: 99%
Catalog No. 50-258-0865 Supplier TARGETMOL CHEMICALS INC Supplier No. T235095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU 0285683 is a specific negative allosteric modulator of mGluR5 with a high affinity for the MPEP binding site. VU 0285683 shows anxiolytic-like activity. purity: 99%