Learn More
Medchemexpress LLC VU 0357121 | 433967-28-3 | C17H17F2NO2 | 1 ML
SDP
Catalog No. 5000376261
Encompass_Preferred
VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or very weakly antagonizing at other mGlu receptor subtypes.
  • Positive and highly selective mGlu5R allosteric modulator (PAM)
  • EC50 of 33 nM for mGlu5 receptor
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Enhances glutamate sensitivity of mGlu5
  • Does not bind at the MPEP allosteric site of mGlu5
  • For research use only
Catalog No. 50-003-76261 Supplier Medchemexpress LLC Supplier No. HY1539310MM/1ML
VU 0357121 is a positive and highly selective mGlu5R allosteric modulator (PAM) with an EC50 of 33 nM. It is inactive or very weakly antagonizing at other mGlu receptor subtypes.
  • Positive and highly selective mGlu5R allosteric modulator (PAM)
  • EC50 of 33 nM for mGlu5 receptor
  • Inactive or very weakly antagonizing at other mGlu receptor subtypes
  • Enhances glutamate sensitivity of mGlu5
  • Does not bind at the MPEP allosteric site of mGlu5
  • For research use only