Learn More
Selleck Chemical LLC VU 0357121 S2795-10mg
SDP
Catalog No. 503112906
Encompass_Preferred
VU0357121 is a novel positive allosteric modulator (PAM) of mGlu5 with EC50 of 33 nM is inactive or very weakly antagonizing at other mGlu receptor subtypes
Catalog No. 50-311-2906 Supplier Selleck Chemical LLC Supplier No. S279510MG
VU0357121 is a novel positive allosteric modulator (PAM) of mGlu5 with EC50 of 33 nM is inactive or very weakly antagonizing at other mGlu receptor subtypes