Learn More
Selleck Chemical LLC VU 0361737 S2892-10mg
SDP
Catalog No. 503111861
Encompass_Preferred
VU 0361737 is a selective positive allosteric modulator (PAM) for mGlu4 receptor with EC50 of 240 nM and 110 nM at human and rat receptors respectively displays weak activity at mGlu5 and mGlu8 receptors is inactive at mGlu1 mGlu2 mGlu3 mGlu6 and mGlu7 receptors can penetrate into CNS
Catalog No. 50-311-1861 Supplier Selleck Chemical LLC Supplier No. S289210MG
VU 0361737 is a selective positive allosteric modulator (PAM) for mGlu4 receptor with EC50 of 240 nM and 110 nM at human and rat receptors respectively displays weak activity at mGlu5 and mGlu8 receptors is inactive at mGlu1 mGlu2 mGlu3 mGlu6 and mGlu7 receptors can penetrate into CNS