Learn More
Medchemexpress LLC VU 0364439 | 1246086-78-1 | C18H13Cl2N3O3S | 1 MG
SDP
Catalog No. 5000376193
Encompass_Preferred
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. It is for research use only. The compound shows better stability in HLM (63% remaining) than RLM (2% remaining). It is a positive allosteric modulator of human mGlu4 receptor expressed in CHO cells, assessed as potentiation of glutamate-induced calcium mobilization. The appearance is a solid, white to off-white.
  • Positive allosteric modulator of human mGlu4 receptor
  • EC50 of 19.8 nM
  • Better stability in HLM (63% remaining) than RLM (2% remaining)
  • Appearance: Solid, white to off-white
  • For research use only
Catalog No. 50-003-76193 Supplier Medchemexpress LLC Supplier No. HY154761MG
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. It is for research use only. The compound shows better stability in HLM (63% remaining) than RLM (2% remaining). It is a positive allosteric modulator of human mGlu4 receptor expressed in CHO cells, assessed as potentiation of glutamate-induced calcium mobilization. The appearance is a solid, white to off-white.
  • Positive allosteric modulator of human mGlu4 receptor
  • EC50 of 19.8 nM
  • Better stability in HLM (63% remaining) than RLM (2% remaining)
  • Appearance: Solid, white to off-white
  • For research use only