Learn More
Medchemexpress LLC VU 0364439 | 1246086-78-1 | C18H13Cl2N3O3S | 100 MG
SDP
Catalog No. 5000376217
Encompass_Preferred
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. While it exhibits less than ideal PK properties for in vivo use, it demonstrates better stability in HLM (63% remaining) compared to RLM (2% remaining).
  • mGlu4 positive allosteric modulator
  • Exhibits an EC50 of 19.8 nM
  • Shows improved stability in HLM (63% remaining) compared to RLM (2% remaining)
  • Soluble in DMSO (50 mg/mL)
  • Targets mGluR in GPCR/G Protein and neuronal signaling pathways
Catalog No. 50-003-76217 Supplier Medchemexpress LLC Supplier No. HY15476100MG
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. While it exhibits less than ideal PK properties for in vivo use, it demonstrates better stability in HLM (63% remaining) compared to RLM (2% remaining).
  • mGlu4 positive allosteric modulator
  • Exhibits an EC50 of 19.8 nM
  • Shows improved stability in HLM (63% remaining) compared to RLM (2% remaining)
  • Soluble in DMSO (50 mg/mL)
  • Targets mGluR in GPCR/G Protein and neuronal signaling pathways