Learn More
Medchemexpress LLC VU 0364439 | 1246086-78-1 | C18H13Cl2N3O3S | 25 MG
SDP
Catalog No. 5000375002
Encompass_Preferred
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. It exhibits better stability in HLM (63% remaining) compared to RLM (2% remaining). It is intended for research use only.
  • mGlu4 positive allosteric modulator (PAM)
  • EC50 of 19.8 nM
  • Better stability in HLM (63% remaining) than RLM (2% remaining)
  • For research use only
Catalog No. 50-003-75002 Supplier Medchemexpress LLC Supplier No. 25MGHY15476
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. It exhibits better stability in HLM (63% remaining) compared to RLM (2% remaining). It is intended for research use only.
  • mGlu4 positive allosteric modulator (PAM)
  • EC50 of 19.8 nM
  • Better stability in HLM (63% remaining) than RLM (2% remaining)
  • For research use only