Learn More
Medchemexpress LLC VU 0364439 | 1246086-78-1 | C18H13Cl2N3O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY154765MG
Encompass_Preferred
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with an EC50 of 19.8 nM. It exhibits better stability in HLM (63% remaining) than RLM (2% remaining), though it possesses less than ideal pharmacokinetic properties preventing its use as an in vivo tool.
  • mGlu4 positive allosteric modulator (PAM)
  • EC50 of 19.8 nM for mGlu4 Receptor
  • High purity of 99.42%
  • White to off-white solid appearance
  • Demonstrates positive allosteric modulation of human mGlu4 receptor in CHO cells
Catalog No. 50-003-76607