Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VU 0364439 | 1246086-78-1 | C18H13Cl2N3O3S | 50 MG
SDP

Catalog No. 5000376908
Encompass_Preferred

VU 0364439 is a mGlu4 positive allosteric modulator (PAM), with an EC50 of 19.8 nM. Intended for research use only, this solid compound appears white to off-white. While it demonstrates better stability in Human Liver Microsomes, its less than ideal PK properties prevent its use as an in vivo tool.

  • Positive allosteric modulator (PAM) for mGlu4
  • EC50 of 19.8 nM
  • Purity of 99.42%
  • Solid appearance, white to off-white color
  • Exhibits better stability in HLM (63% remaining) compared to RLM (2% remaining)
  • Target: mGluR

Catalog No. 50-003-76908 Supplier Medchemexpress LLC Supplier No. HY1547650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU 0364439 is a mGlu4 positive allosteric modulator (PAM), with an EC50 of 19.8 nM. Intended for research use only, this solid compound appears white to off-white. While it demonstrates better stability in Human Liver Microsomes, its less than ideal PK properties prevent its use as an in vivo tool.

  • Positive allosteric modulator (PAM) for mGlu4
  • EC50 of 19.8 nM
  • Purity of 99.42%
  • Solid appearance, white to off-white color
  • Exhibits better stability in HLM (63% remaining) compared to RLM (2% remaining)
  • Target: mGluR

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.