Learn More
Selleck Chemical LLC VU 0364439
SDP
Catalog No. 5000000202
Encompass_Preferred
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with EC50 of 19 8 nM
Catalog No. 50-000-00202 Supplier Selleck Chemical LLC Supplier No. S803510MG
VU 0364439 is a mGlu4 positive allosteric modulator (PAM) with EC50 of 19 8 nM