Learn More
Medchemexpress LLC Vu 0365114 | 1208222-39-2 | 99.4% | 397.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY107651
Encompass_Preferred
VU 0365114 is a selective mAChR M5 positive allosteric modulator, with an EC50 of 2.7 μM, and >30 μM for M1, M2, M3 and M4 receptors. VU 0365114 increases insulin secretion stimulated by ACh in human β-cells.
  • Selective mAChR M5 positive allosteric modulator
  • Increases insulin secretion stimulated by ACh in human β-cells
Catalog No. 50-003-65938