Learn More
Medchemexpress LLC HY-107651 5mg Medchemexpress, VU 0365114 CAS:1208222-39-2 Purity:>98%
SDP
Catalog No. 501968449
Encompass_Preferred
Medchemexpress, HY-107651 5mg VU 0365114 CAS:1208222-39-2 VU 0365114 is a mAChR M5 positive allosteric modulator, with an EC50 of 2.7 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8449 Supplier Medchemexpress LLC Supplier No. HY1076515MG
Medchemexpress, HY-107651 5mg VU 0365114 CAS:1208222-39-2 VU 0365114 is a mAChR M5 positive allosteric modulator, with an EC50 of 2.7 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.