Learn More
Selleck Chemical LLC VU 0365114-E0799-5MG
SDP
Catalog No. 503135315
Encompass_Preferred
VU 0365114 is a muscarinic acetylcholine receptor M5-preferring (mAChR M5-preferring) positive allosteric modulator (PAM) with an EC50 of 2 7 M
Catalog No. 50-313-5315 Supplier Selleck Chemical LLC Supplier No. E07995MG
VU 0365114 is a muscarinic acetylcholine receptor M5-preferring (mAChR M5-preferring) positive allosteric modulator (PAM) with an EC50 of 2 7 M