Learn More
Medchemexpress LLC VU-1545 | 890764-63-3 | 99.0% | 402.38 | 100 MG
SDP
Catalog No. 5000391134
Encompass_Preferred
VU-1545 is a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM) with a Ki of 156 nM and an EC50 of 9.6 nM.
  • Promotes AKT activation at concentrations of 0.1 and 1.0 μM.
  • Neuroprotective in a mouse model of Huntington's disease.
  • For research use only, not for sale to patients.
Catalog No. 50-003-91134 Supplier Medchemexpress LLC Supplier No. HY16951100MG
VU-1545 is a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM) with a Ki of 156 nM and an EC50 of 9.6 nM.
  • Promotes AKT activation at concentrations of 0.1 and 1.0 μM.
  • Neuroprotective in a mouse model of Huntington's disease.
  • For research use only, not for sale to patients.