Learn More
Medchemexpress LLC VU-1545 | 890764-63-3 | 99.0% | 402.38 | 25 MG
SDP
Catalog No. 5000394912
Encompass_Preferred
VU-1545 is a potent metabotropic glutamate receptor 5 (mGluR5) positive allosteric modulator (PAM) with a Ki of 156 nM and an EC50 of 9.6 nM. It promotes AKT activation, efficiently doing so at concentrations as low as 0.1 and 1.0 μM. Studies have shown that VU-1545 at 10 and 100 μM promotes Akt phosphorylation. This compound is used for research purposes and is not intended for patient use.
  • Potent mGluR5 positive allosteric modulator
  • Ki of 156 nM for mGluR5
  • EC50 of 9.6 nM for mGluR5
  • Promotes AKT activation in neuronal cells
  • Efficiently promotes Akt phosphorylation at 10.0 μM and 100.0 μM
Catalog No. 50-003-94912 Supplier Medchemexpress LLC Supplier No. 25MGHY16951
VU-1545 is a potent metabotropic glutamate receptor 5 (mGluR5) positive allosteric modulator (PAM) with a Ki of 156 nM and an EC50 of 9.6 nM. It promotes AKT activation, efficiently doing so at concentrations as low as 0.1 and 1.0 μM. Studies have shown that VU-1545 at 10 and 100 μM promotes Akt phosphorylation. This compound is used for research purposes and is not intended for patient use.
  • Potent mGluR5 positive allosteric modulator
  • Ki of 156 nM for mGluR5
  • EC50 of 9.6 nM for mGluR5
  • Promotes AKT activation in neuronal cells
  • Efficiently promotes Akt phosphorylation at 10.0 μM and 100.0 μM