Learn More
Medchemexpress LLC VU-1545 | 890764-63-3 | 99.0% | 402.38 | 50 MG
SDP
Catalog No. 5000390599
Encompass_Preferred
VU-1545 is a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM). It exhibits a Ki of 156 nM and an EC50 of 9.6 nM. This compound promotes AKT activation at concentrations of 0.1 and 1.0 μM and is intended for research use only.
  • Functions as a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM).
  • Demonstrates a Ki of 156 nM and an EC50 of 9.6 nM.
  • Promotes AKT activation.
  • Promotes Akt phosphorylation in neuronal cells.
  • Solid appearance, light yellow to yellow color.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-003-90599 Supplier Medchemexpress LLC Supplier No. HY1695150MG
VU-1545 is a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM). It exhibits a Ki of 156 nM and an EC50 of 9.6 nM. This compound promotes AKT activation at concentrations of 0.1 and 1.0 μM and is intended for research use only.
  • Functions as a metabotropic glutamate receptor 5 positive allosteric modulator (mGluR5 PAM).
  • Demonstrates a Ki of 156 nM and an EC50 of 9.6 nM.
  • Promotes AKT activation.
  • Promotes Akt phosphorylation in neuronal cells.
  • Solid appearance, light yellow to yellow color.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.