Learn More
Selleck Chemical LLC VU-29 S1970-25mg
SDP
Catalog No. 503109600
Encompass_Preferred
VU-29 is a positive allosteric modulator of metabotropic glutamate 5 (mGlu5) receptor with EC50 of 9 nM and Ki 244 nM for rat mGluR5 which is selective for mGluR5 relative to other mGluR subtypes
Catalog No. 50-310-9600 Supplier Selleck Chemical LLC Supplier No. S197025MG
VU-29 is a positive allosteric modulator of metabotropic glutamate 5 (mGlu5) receptor with EC50 of 9 nM and Ki 244 nM for rat mGluR5 which is selective for mGluR5 relative to other mGluR subtypes