Learn More
Abcam VU0119498, M1 muscarinic receptor agonist and pan mAChR M3, M5 PAM, 10MG
Catalog No. 502706541
Encompass_Preferred
MW 316.15 Da, Purity >99%. M1 muscarinic receptor agonist (EC50 = 3.1 μM) and pan mAChR M3, M5 positive allosteric modulator (PAM). Neuroprotective agent. Active *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6541 Supplier Abcam Supplier No. AB14151210MG
MW 316.15 Da, Purity >99%. M1 muscarinic receptor agonist (EC50 = 3.1 μM) and pan mAChR M3, M5 positive allosteric modulator (PAM). Neuroprotective agent. Active *in vitro*.
The product is subject to the following: Abcam Restricted Use Statement