Learn More
Medchemexpress LLC Vu0134992 hydrochloride | 1052515-91-9 | 98.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY122560A10MM 1ML
Encompass_Preferred
VU0134992 hydrochloride is a selective Kir4.1 inward-rectifier potassium channel blocker provided as the hydrochloride salt. Supplied as a 10 mM solution in DMSO or as a solid, it is used as a research tool to inhibit Kir4.1-mediated currents in electrophysiology and cell-based assays.
  • Selective Kir4.1 blocker (IC50 = 0.97 μM).
  • Supplied as hydrochloride salt for improved solubility.
  • Available as 10 mM solution in DMSO or in solid form.
  • Suitable for electrophysiology and cell-based assays.
  • High purity suitable for research applications.
Catalog No. 50-002-30247