Learn More
Abcam VU0155041, positive allosteric mGluR4 agonist, 10MG
Catalog No. 502707754
Encompass_Preferred
MW 316.2 Da, Purity >99%. Positive allosteric selective modulator at mGluR4 (EC50 values are 798 and 693 nM at human and rat mGluR4 receptors, respectively). It has advantages over PHCCC (ab120043) as it is more potent. Shows limited blood-brain barrier permeability. Active in behavioural models of Parkinson's disease following icv injection. Available in water-soluble form - please see VU0155041 sodium salt (ab120248).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7754 Supplier Abcam Supplier No. AB12024410MG
MW 316.2 Da, Purity >99%. Positive allosteric selective modulator at mGluR4 (EC50 values are 798 and 693 nM at human and rat mGluR4 receptors, respectively). It has advantages over PHCCC (ab120043) as it is more potent. Shows limited blood-brain barrier permeability. Active in behavioural models of Parkinson's disease following icv injection. Available in water-soluble form - please see VU0155041 sodium salt (ab120248).
The product is subject to the following: Abcam Restricted Use Statement