Learn More
TARGETMOL CHEMICALS INC VU0155069 5MG
SDP
Catalog No. 502581111
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU0155069 strongly inhibits the invasive migration of several cancer cell lines in transwell assays. VU0155069 is a selective phospholipase D1 inhibitor (IC50 46 nM in vitro). purity: 98%
Catalog No. 50-258-1111 Supplier TARGETMOL CHEMICALS INC Supplier No. T172395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VU0155069 strongly inhibits the invasive migration of several cancer cell lines in transwell assays. VU0155069 is a selective phospholipase D1 inhibitor (IC50 46 nM in vitro). purity: 98%