Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vu0238441 | 85511-68-8 | 99.9% | 10 MG
SDP

Catalog No. 5000234778
Encompass_Preferred

VU0238441 is a pan muscarinic acetylcholine receptor (mAChR) positive allosteric modulator used as a research tool to potentiate M1-M5 receptor responses in pharmacology and neuroscience studies.

  • Potentiates M1-M5 receptor responses; EC50s: M1 3.2 μM, M2 2.8 μM, M3 2.2 μM, M5 2.1 μM, M4 >10 μM.
  • High purity (99.9%).
  • Molecular formula C16H9ClF3NO2; molecular weight 339.70 g·mol⁻¹.
  • Available as powder and as a 10 mM solution in DMSO; common pack sizes from 5 mg to 500 mg.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-34778 Supplier Medchemexpress LLC Supplier No. HY1215810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0238441 is a pan muscarinic acetylcholine receptor (mAChR) positive allosteric modulator used as a research tool to potentiate M1-M5 receptor responses in pharmacology and neuroscience studies.

  • Potentiates M1-M5 receptor responses; EC50s: M1 3.2 μM, M2 2.8 μM, M3 2.2 μM, M5 2.1 μM, M4 >10 μM.
  • High purity (99.9%).
  • Molecular formula C16H9ClF3NO2; molecular weight 339.70 g·mol⁻¹.
  • Available as powder and as a 10 mM solution in DMSO; common pack sizes from 5 mg to 500 mg.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.