Learn More
Medchemexpress LLC VU0357017 hydrochloride | 1135242-13-5 | 98.63% | 369.89 | 1 ML
SDP
Catalog No. 5000402557
Encompass_Preferred
VU0357017 hydrochloride is a potent, selective, and brain-penetrant allosteric agonist of the M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. It is highly selective for M1, showing no activity at M2-M5 receptors up to 30 μM. This compound is used in research for conditions such as Alzheimer's disease and schizophrenia.
  • Potent and selective M1 muscarinic acetylcholine receptor agonist.
  • Exhibits brain penetrant properties.
  • Highly selective for M1 over M2-M5 receptors.
  • Used for the research of Alzheimer's disease and schizophrenia.
Catalog No. 50-004-02557 Supplier Medchemexpress LLC Supplier No. HY19752A10MM/1ML
VU0357017 hydrochloride is a potent, selective, and brain-penetrant allosteric agonist of the M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. It is highly selective for M1, showing no activity at M2-M5 receptors up to 30 μM. This compound is used in research for conditions such as Alzheimer's disease and schizophrenia.
  • Potent and selective M1 muscarinic acetylcholine receptor agonist.
  • Exhibits brain penetrant properties.
  • Highly selective for M1 over M2-M5 receptors.
  • Used for the research of Alzheimer's disease and schizophrenia.