Learn More
Medchemexpress LLC VU0357017 hydrochloride | 1135242-13-5 | MFCD17215959 | 99.9% | 369.89 | C18H28ClN3O3 | 10 MG
SDP
Catalog No. 5000298346
Encompass_Preferred
VU0357017 hydrochloride is a brain-penetrant, selective allosteric agonist of the M1 muscarinic acetylcholine receptor used as a research tool to probe M1 receptor pharmacology and signaling. Supplied as the hydrochloride salt, the compound is characterized by a defined molecular weight and high reported purity.
  • Potent M1 allosteric agonist with reported EC50 ≈ 477 nM.
  • Brain-penetrant small molecule suitable for in vitro and in vivo studies.
  • High chemical purity as provided by manufacturer.
  • Provided as a hydrochloride salt for improved solubility.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-98346 Supplier Medchemexpress LLC Supplier No. HY19752A10MG
VU0357017 hydrochloride is a brain-penetrant, selective allosteric agonist of the M1 muscarinic acetylcholine receptor used as a research tool to probe M1 receptor pharmacology and signaling. Supplied as the hydrochloride salt, the compound is characterized by a defined molecular weight and high reported purity.
  • Potent M1 allosteric agonist with reported EC50 ≈ 477 nM.
  • Brain-penetrant small molecule suitable for in vitro and in vivo studies.
  • High chemical purity as provided by manufacturer.
  • Provided as a hydrochloride salt for improved solubility.
  • Available in small research pack sizes for laboratory use.