Learn More
Selleck Chemical LLC VU0357017 Hydrochloride
SDP
Catalog No. 5000002325
Encompass_Preferred
VU0357017 hydrochloride (CID-25010775) is a potent highly selective and CNS-penetrant agonist of M1 which is a subtype of muscarinic acetylcholine receptors (mAChRs) VU0357017 hydrochloride appears to act at an allosteric site to activate the receptor with EC50 of 477 nM and Ki of 9 91 M
Catalog No. 50-000-02325 Supplier Selleck Chemical LLC Supplier No. S67955MG
VU0357017 hydrochloride (CID-25010775) is a potent highly selective and CNS-penetrant agonist of M1 which is a subtype of muscarinic acetylcholine receptors (mAChRs) VU0357017 hydrochloride appears to act at an allosteric site to activate the receptor with EC50 of 477 nM and Ki of 9 91 M