Learn More
Medchemexpress LLC HY-19752A 5mg Medchemexpress, VU0357017 (hydrochloride) CAS:1135242-13-5 Purity:>98%
SDP
Catalog No. 502082176
Encompass_Preferred
Medchemexpress, HY-19752A 5mg VU0357017 (hydrochloride) CAS:1135242-13-5 VU0357017 hydrochloride (CID-25010775) is a potent, selective and brain-penetrant allosteric agonist of M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. VU0357017 hydrochloride is highly selective for M1 and has no activity at M2-M5 up to the highest concentrations tested (30 μM). VU0357017 hydrochloride can be used for the research of Alzheimer’s disease and schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2176 Supplier Medchemexpress LLC Supplier No. HY19752A5MG
Medchemexpress, HY-19752A 5mg VU0357017 (hydrochloride) CAS:1135242-13-5 VU0357017 hydrochloride (CID-25010775) is a potent, selective and brain-penetrant allosteric agonist of M1 muscarinic acetylcholine receptor, with an EC50 of 477 nM. VU0357017 hydrochloride is highly selective for M1 and has no activity at M2-M5 up to the highest concentrations tested (30 μM). VU0357017 hydrochloride can be used for the research of Alzheimer’s disease and schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.