Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vu0360172 | 1310012-12-4 | 98.1% | 5 MG
SDP

Catalog No. 5000232205
Encompass_Preferred

Small-molecule positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) supplied for research use in small milligram quantities.

  • High purity (98.1%).
  • Molecular weight 294.32 g/mol.
  • Molecular formula C18H15FN2O.
  • Suitable for in vitro pharmacology and receptor modulation studies.
  • Available in small milligram sizes for assay development.

Catalog No. 50-002-32205 Supplier Medchemexpress LLC Supplier No. HY1205895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) supplied for research use in small milligram quantities.

  • High purity (98.1%).
  • Molecular weight 294.32 g/mol.
  • Molecular formula C18H15FN2O.
  • Suitable for in vitro pharmacology and receptor modulation studies.
  • Available in small milligram sizes for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.