Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VU0360172 hydrochloride | 1309976-62-2 | 98.9% | 330.78 | C18H16ClFN2O | 5 MG
SDP

Catalog No. 5000240803
Encompass_Preferred

VU0360172 hydrochloride is a potent, selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 5 (mGlu5). It potentiates mGlu5 signaling (EC50 = 16 nM; Ki = 195 nM) and is supplied as a white to off-white solid for research use. Typical applications include receptor pharmacology, cellular signaling assays, and in vivo pharmacology studies.

  • Potent mGlu5 PAM activity (EC50 16 nM).
  • Confirmed binding affinity (Ki 195 nM).
  • High purity suitable for research (98.9%).
  • Molecular weight 330.78 for accurate dosing and calculations.
  • Solid form, white to off-white, for convenient handling and storage.
  • Recommended storage: store sealed at -20°C; in solution, -80°C for long term.
  • Supplied in small research pack sizes appropriate for pilot studies.

Catalog No. 50-002-40803 Supplier Medchemexpress LLC Supplier No. HY1075045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0360172 hydrochloride is a potent, selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 5 (mGlu5). It potentiates mGlu5 signaling (EC50 = 16 nM; Ki = 195 nM) and is supplied as a white to off-white solid for research use. Typical applications include receptor pharmacology, cellular signaling assays, and in vivo pharmacology studies.

  • Potent mGlu5 PAM activity (EC50 16 nM).
  • Confirmed binding affinity (Ki 195 nM).
  • High purity suitable for research (98.9%).
  • Molecular weight 330.78 for accurate dosing and calculations.
  • Solid form, white to off-white, for convenient handling and storage.
  • Recommended storage: store sealed at -20°C; in solution, -80°C for long term.
  • Supplied in small research pack sizes appropriate for pilot studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.