Learn More
Medchemexpress LLC VU0360172 hydrochloride | 1309976-62-2 | 98.9% | 330.78 g/mol | C18H16ClFN2O | 100 MG
SDP
Catalog No. 5000240622
Encompass_Preferred
VU0360172 hydrochloride is a potent and selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 5 (mGlu5), used in pharmacology and neuroscience research to probe mGlu5 signaling. Reported values include EC50 16 nM and Ki 195 nM. The compound is supplied as a white to off-white solid with high purity and recommended cold storage for stability.
  • Potent mGlu5 activity (EC50 16 nM).
  • Selective binding profile (Ki 195 nM for mGlu5).
  • High purity suitable for research use (approximately 98.9%).
  • Solid, white to off-white physical form for easy handling.
  • Stable when stored cold (recommended -20°C; in solvent: -80°C for extended storage).
  • Available in multiple small pack sizes for research workflows.
Catalog No. 50-002-40622 Supplier Medchemexpress LLC Supplier No. HY107504100MG
VU0360172 hydrochloride is a potent and selective positive allosteric modulator (PAM) of the metabotropic glutamate receptor 5 (mGlu5), used in pharmacology and neuroscience research to probe mGlu5 signaling. Reported values include EC50 16 nM and Ki 195 nM. The compound is supplied as a white to off-white solid with high purity and recommended cold storage for stability.
  • Potent mGlu5 activity (EC50 16 nM).
  • Selective binding profile (Ki 195 nM for mGlu5).
  • High purity suitable for research use (approximately 98.9%).
  • Solid, white to off-white physical form for easy handling.
  • Stable when stored cold (recommended -20°C; in solvent: -80°C for extended storage).
  • Available in multiple small pack sizes for research workflows.