Learn More
Medchemexpress LLC VU0360172 hydrochloride | 1309976-62-2 | 98.9% | 330.78 g/mol | C18H16ClFN2O | 25 MG
SDP
Catalog No. 5000240621
Encompass_Preferred
VU0360172 hydrochloride is a potent and selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used as a research tool in pharmacology and neuroscience. Supplied as the hydrochloride salt, the compound is intended for in vitro assays and preclinical in vivo studies and is provided with high purity and defined storage recommendations.
  • Potent mGlu5 positive allosteric modulator (EC50 = 16 nM).
  • Selective pharmacological tool for mGlu5 receptor studies.
  • High purity suitable for biochemical and preclinical assays.
  • Multiple milligram pack sizes available for laboratory experiments.
  • Recommended storage conditions provided to maintain stability.
Catalog No. 50-002-40621 Supplier Medchemexpress LLC Supplier No. HY10750425MG
VU0360172 hydrochloride is a potent and selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used as a research tool in pharmacology and neuroscience. Supplied as the hydrochloride salt, the compound is intended for in vitro assays and preclinical in vivo studies and is provided with high purity and defined storage recommendations.
  • Potent mGlu5 positive allosteric modulator (EC50 = 16 nM).
  • Selective pharmacological tool for mGlu5 receptor studies.
  • High purity suitable for biochemical and preclinical assays.
  • Multiple milligram pack sizes available for laboratory experiments.
  • Recommended storage conditions provided to maintain stability.