Learn More
Medchemexpress LLC HY-100588 5mg Medchemexpress, VU0364770 CAS:61350-00-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1005885MG
Encompass_Preferred
Medchemexpress, HY-100588 5mg VU0364770 CAS:61350-00-3 VU0364770 is a selective and potent positive allosteric modulator ( PAM ) of mGlu4 . VU0346770 exhibits EC 50 s of 290 nM and 1.1 μM at rat mGlu4 and human mGlu4 receptor , respectively. VU0364770 exhibits antagonist activity at mGlu5 with a potency of 17.9 μM and PAM activity at mGlu6 with a potency of 6.8 μM. VU0364770 also possesses activity at MAO with K i values of 8.5 and 0.72 μM for human MAO-A and human MAO-B, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1895