Learn More
Medchemexpress LLC HY-100588A 10mg Medchemexpress, VU0364770 (hydrochloride) CAS:1414842-70-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100588A10MG
Encompass_Preferred
Medchemexpress, HY-100588A 10mg VU0364770 (HCl) CAS:1414842-70-8 VU0364770 hydrochloride is a selective and potent positive allosteric modulator (PAM) of mGlu4. VU0346770 hydrochloride exhibits EC50s of 290 nM and 1.1 μM at rat mGlu4 and human mGlu4 receptor, respectively. VU0364770 hydrochloride exhibits antagonist activity at mGlu5 with a potency of 17.9 μM and PAM activity at mGlu6 with a potency of 6.8 μM. VU0364770 hydrochloride also possesses activity at MAO with Ki values of 8.5 and 0.72 μM for human MAO-A and human MAO-B, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7751